missing translation for 'onlineSavingsMsg'
Learn More

2-Methylfuran 98.0+%, TCI America™

Catalog No. M0226500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0226500ML 500 mL
M022625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0226500ML Supplier TCI America Supplier No. M0226500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 534-22-5
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.102
MDL Number MFCD00003248
InChI Key VQKFNUFAXTZWDK-UHFFFAOYSA-N
Synonym methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
PubChem CID 10797
IUPAC Name 2-methylfuran
SMILES CC1=CC=CO1

Specifications

Color Yellow
Boiling Point 63°C
Quantity 500 mL
UN Number 2301
Formula Weight 82.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylfuran
EINECSNumber (5)-5544
RTECSNumber LU2625000
TSCA Yes