missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylfuran, 98+%, stab.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -89°C |
| Density | 0.91 |
| Boiling Point | 63°C to 65°C |
| Flash Point | -30°C (-22°F) |
| Assay Percent Range | 98+% |
| Refractive Index | 1.433 |
| Quantity | 1000 mL |
| UN Number | UN2301 |
| Beilstein | 103733 |
| Formula Weight | 82.1 |
| Show More |
Safety and Handling
| Hazard Category | H225-H301+H311+H331 |
| Hazard Statement | GHS H StatementH225-H300-H330-H341-H319Highly flammable liquid and vapour.Fatal if swallowed.Fatal if inhaled.Suspected of causing genetic defects.Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-METHYLFURAN |
| EINECSNumber | 208-594-5 |
| RTECSNumber | LU2625000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only