missing translation for 'onlineSavingsMsg'
Learn More

Cesium carbonate, Puratronic™, 99.994% (metals basis)

Catalog No. AA1211722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1211722 100 g
AA1211706 5 g
AA1211714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1211722 Supplier Thermo Scientific Chemicals Supplier No. 01211722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 534-17-8 | CCs2O3 | 325.82 g/mol

Stainless Steel gauze is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O

Specifications

Melting Point 610°C (decomposition)
Color White
Physical Form Powder/Granules
Quantity 100 g
Assay Percent Range 99.994% (Metal basis)
Beilstein 4546405
Solubility Information Highly soluble in water. Soluble in alcohol,dimethyl sulfoxide and N,N-dimethylformamide. Insoluble in toluene and p-xylene.
IUPAC Name dicaesium(1+) carbonate
Formula Weight 325.82
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 4.072 g/cm3 at 20°C
Chemical Name or Material Cesium carbonate
Show More Show Less
Hazard Category H318-H361f-H373
Precautionary Statement P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c
EINECSNumber 208-591-9
RTECSNumber FK9400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only