accessibility menu, dialog, popup

UserName

H2NOHHO

CAS RN 534-03-2

IUPAC Name: 2-aminopropane-1,3-diol
Synonyms: 2-aminopropane-1,3-diol
Molecular Weight (g/mol): 91.11
Molecular Formula: C3H9NO2
InChi Key: KJJPLEZQSCZCKE-UHFFFAOYSA-N
SMILES: NC(CO)CO
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)