missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3-propanediol, 98%

Catalog No. AAL0842614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0842614 25 g
AAL0842606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0842614 Supplier Thermo Scientific Chemicals Supplier No. L0842614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 534-03-2 | C3H9NO2 | 91.11 g/mol

2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 534-03-2
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00051487
InChI Key KJJPLEZQSCZCKE-UHFFFAOYSA-N
Synonym 2-amino-1,3-propanediol, serinol, 1,3-propanediol, 2-amino, 2-amino-1,3-propandiol, 2-aminopropan-1,3-diol, unii-ic94l30j8m, 2-amino-1,3-dihydroxypropane, 2-amino-1,3-propanediol serinol, dl-serinol, sel
PubChem CID 68294
IUPAC Name 2-aminopropane-1,3-diol
SMILES NC(CO)CO

Specifications

Melting Point 52°C to 56°C
Boiling Point 128°C (0.8 mmHg)
Quantity 25 g
UN Number UN3259
Beilstein 635683
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water and alcohol.
Formula Weight 91.11
Percent Purity 98%
Chemical Name or Material 2-Amino-1,3-propanediol
Hazard Category H315-H318
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 208-584-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only