accessibility menu, dialog, popup

UserName

H3COOCH3CH2

CAS RN 53399-81-8

IUPAC Name: ethyl 2-methylpent-4-enoate
Synonyms: ethyl 2-methylpent-4-enoate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: BDBGKYIBDXAVMX-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)CC=C
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)