missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methyl-4-pentenoate, 98%

Catalog No. AAA1952622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1952622 100 g
AAA1952606 5 g
AAA1952614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1952622 Supplier Thermo Scientific Chemicals Supplier No. A1952622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53399-81-8 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53399-81-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00010235
InChI Key BDBGKYIBDXAVMX-UHFFFAOYNA-N
Synonym ethyl 2-methyl-4-pentenoate, 2-methyl-4-pentenoic acid ethyl ester, ethyl 2-methyl 4-pentenoate, 4-pentenoic acid, 2-methyl-, ethyl ester, ethyl 2-methylpent-4-en-1-oate, ethyl 2-methyl-4-pentanoate, fema no. 3489, pineapple pentenoate, acmc-1ao14, ethyl-2-methyl-4-pentenoate
PubChem CID 62024
IUPAC Name ethyl 2-methylpent-4-enoate
SMILES CCOC(=O)C(C)CC=C

Specifications

Density 0.88
Boiling Point 153°C to 155°C
Flash Point 40°C (104°F)
Refractive Index 1.416
Quantity 100 g
UN Number UN3272
Beilstein 1753406
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Ethyl 2-methyl-4-pentenoate
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 258-520-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only