missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Methyl-4-pentenoate 98.0+%, TCI America™

Catalog No. E07175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07175G Supplier TCI America Supplier No. E07175G

Chemical Identifiers

CAS 53399-81-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00010235
InChI Key BDBGKYIBDXAVMX-UHFFFAOYNA-N
Synonym ethyl 2-methyl-4-pentenoate, 2-methyl-4-pentenoic acid ethyl ester, ethyl 2-methyl 4-pentenoate, 4-pentenoic acid, 2-methyl-, ethyl ester, ethyl 2-methylpent-4-en-1-oate, ethyl 2-methyl-4-pentanoate, fema no. 3489, pineapple pentenoate, acmc-1ao14, ethyl-2-methyl-4-pentenoate
PubChem CID 62024
IUPAC Name ethyl 2-methylpent-4-enoate
SMILES CCOC(=O)C(C)CC=C

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 g
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Methyl-4-pentenoate