missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium 2-naphthalenesulfonate , ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014491
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014491 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014491 Supplier Merck Emd Millipore Supplier No. 7028910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory-grade naphthalene-2-sulfonic acid sodium salt suitable for chemical research and manufacturing applications.

Chemical Identifiers

CAS 532-02-5
Molecular Weight (g/mol) 230.22
MDL Number MFCD00064186
Synonym 2-Naphthalenesulfonic acid sodium salt

Specifications

Physical Form Powder
Quantity 10 g
Linear Formula C10H7 NaO3 S
Formula Weight 230.22
Percent Purity ≥99.0% (HPLC)
Grade Ion Pair Chromatography
Product Line LiChropur
Chemical Name or Material Sodium Naphthalene-2-sulfonate
RTECSNumber QK3678000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany