accessibility menu, dialog, popup

UserName

HNHN

CAS RN 531-91-9

IUPAC Name: N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 336.44
Molecular Formula: C24H20N2
InChi Key: FDRNXKXKFNHNCA-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)