accessibility menu, dialog, popup

UserName

ClClP

CAS RN 5293-84-5

IUPAC Name: (chloromethyl)triphenylphosphanium chloride
Synonyms: (chloromethyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 347.22
Molecular Formula: C19H17Cl2P
InChi Key: SXYFAZGVNNYGJQ-UHFFFAOYSA-M
SMILES: [Cl-].ClC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results