missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)triphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. C10095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10095G 5 g
C100925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10095G Supplier TCI America Supplier No. C10095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5293-84-5
Molecular Formula C19H17Cl2P
Molecular Weight (g/mol) 347.22
MDL Number MFCD00011867
InChI Key SXYFAZGVNNYGJQ-UHFFFAOYSA-M
Synonym chloromethyl triphenylphosphonium chloride, chloromethyltriphenylphosphonium chloride, chloromethyl triphenylphosphoniumchloride, phosphonium, chloromethyltriphenyl-, chloride, chloromethly triphenylphosphonium chloride, chloromethyl triphenylphosphanium chloride, chloromethyl triphenylphosphonium chliride, triphenyl chloromethyl-phosphonium chloride, acmc-1albk, phosphonium, chloromethyl triphenyl-, chloride
PubChem CID 197823
IUPAC Name (chloromethyl)triphenylphosphanium chloride
SMILES [Cl-].ClC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 347.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Chloromethyl)triphenylphosphonium Chloride
RTECSNumber TA1882000
TSCA No