accessibility menu, dialog, popup

UserName

H3CON

CAS RN 52709-86-1

IUPAC Name: 4'-propoxy-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-propoxy-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 237.3
Molecular Formula: C16H15NO
InChi Key: RZCQJZFWJZHMPK-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results