CAS RN 527-84-4
CAS RN 527-84-4
IUPAC Name:
1-methyl-2-(propan-2-yl)benzene
Synonyms:
1-methyl-2-isopropylbenzene
Molecular Weight (g/mol):
134.22
Molecular Formula:
C10H14
InChi Key:
WWRCMNKATXZARA-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC=C1C
1
–
1
of
1
results
1
o-Cymene 99.0+%, TCI America™
CAS: 527-84-4 Molecular Formula: C10H14 Molecular Weight (g/mol): 134.222 MDL Number: MFCD00008888 InChI Key: WWRCMNKATXZARA-UHFFFAOYSA-N Synonym: 2-Isopropyltoluene, 2-Methylisopropylbenzene PubChem CID: 10703 IUPAC Name: 1-methyl-2-propan-2-ylbenzene SMILES: CC1=CC=CC=C1C(C)C