missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Cymene 99.0+%, TCI America™
Specifications
Specifications
| Physical Form | Liquid |
| UN Number | 2046 |
| Color | Colorless |
| Boiling Point | 178°C |
| Quantity | 0.1 mL |
| Formula Weight | 134.22 |
| Percent Purity | ≥99.0% (GC) |
| Chemical Name or Material | o-Cymene |
Safety and Handling
| EINECSNumber | (3)-0012&(3)-0015 |
| RTECSNumber | DA6127300 |
| TSCA | Yes |