accessibility menu, dialog, popup

UserName

NH(R)HNOHOOCH3CH3CH3O

CAS RN 5241-64-5

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(1H-indol-3-yl)propanoic acid|BOC-D-TRP-OH
Molecular Weight (g/mol): 304.35
Molecular Formula: C16H20N2O4
InChi Key: NFVNYBJCJGKVQK-CYBMUJFWSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results