CAS RN 5197-87-5
IUPAC Name:
benzyltripropylazanium chloride
Synonyms:
benzyltripropylazanium chloride
Molecular Weight (g/mol):
269.86
Molecular Formula:
C16H28ClN
InChi Key:
YTRIOKYQEVFKGU-UHFFFAOYSA-M
SMILES:
[Cl-].CCC[N+](CCC)(CCC)CC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Benzyltri-n-propylammonium chloride, 96%
Benzyltri-n-propylammonium chloride, 96%
CAS: 5197-87-5 Molecular Formula: C16H28ClN Molecular Weight (g/mol): 269.86 MDL Number: MFCD00051869 InChI Key: YTRIOKYQEVFKGU-UHFFFAOYSA-M Synonym: benzyltripropylammonium chloride,n-benzyl-n,n-dipropylpropan-1-aminium chloride,benzyl tripropyl ammonium chloride,benzyltri-n-propylammonium chloride,benzyltripropylazanium chloride,benzyl tripropyl azanium chloride,acmc-20alov,c16h28n.cl,benzyl-tri-n-propylammonium chloride PubChem CID: 6097067 IUPAC Name: benzyltripropylazanium chloride SMILES: [Cl-].CCC[N+](CCC)(CCC)CC1=CC=CC=C1