CAS RN 51516-68-8
IUPAC Name:
5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
Synonyms:
5-amino-1-(3-chlorophenyl)pyrazole-4-carbonitrile
Molecular Weight (g/mol):
218.64
Molecular Formula:
C10H7ClN4
InChi Key:
VRKRSWGTAMOYEV-UHFFFAOYSA-N
SMILES:
NC1=C(C=NN1C1=CC=CC(Cl)=C1)C#N
1
–
1
of
1
results
1
Promotions Available
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%
CAS: 51516-68-8 Molecular Formula: C10H7ClN4 Molecular Weight (g/mol): 218.64 MDL Number: MFCD00128298 InChI Key: VRKRSWGTAMOYEV-UHFFFAOYSA-N Synonym: 5-amino-1-3-chlorophenyl-1h-pyrazole-4-carbonitrile,5-amino-1-3-chlorophenyl pyrazole-4-carbonitrile,5-amino-1-3-chlorophenyl-1h-pyrazole-,5-amino-1-3-chloro-phenyl-1h-pyrazole-4-carbonit,1h-pyrazole-4-carbonitrile,5-amino-1-3-chlorophenyl,1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl PubChem CID: 734496 IUPAC Name: 5-amino-1-(3-chlorophenyl)pyrazole-4-carbonitrile SMILES: NC1=C(C=NN1C1=CC=CC(Cl)=C1)C#N