accessibility menu, dialog, popup

UserName

CAS RN 515-69-5

CH3CH3(S)H3COH(R)CH3

CAS RN 515-69-5

IUPAC Name: (2S)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
Synonyms: (2S)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
Molecular Weight (g/mol): 222.37
Molecular Formula: C15H26O
InChi Key: RGZSQWQPBWRIAQ-GJZGRUSLSA-N
SMILES: CC(C)=CCC[C@](C)(O)[C@@H]1CCC(C)=CC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)