missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-alpha-Bisabolol 80.0+%, TCI America™

Catalog No. B211925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B211925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. B211925ML missing translation for 'mfr' TCI America missing translation for 'supplierNo' B211925ML

Chemical Identifiers

CAS 515-69-5
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.372
MDL Number MFCD00129082
InChI Key RGZSQWQPBWRIAQ-LSDHHAIUSA-N
Synonym bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260
PubChem CID 1549992
IUPAC Name (2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
SMILES CC1=CCC(CC1)C(C)(CCC=C(C)C)O

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 222.37
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material (+/-)-alpha-Bisabolol
RTECSNumber MJ9685000
TSCA Yes