missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bisabolol, 96%

Catalog No. AAB2145718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2145718 50 g
AAB2145709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2145718 Supplier Thermo Scientific Chemicals Supplier No. B2145718

CAS: 515-69-5 | C15H26O | 222.372 g/mol

It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

Solubility
Soluble in alcohol, natural, mineral and synthetic oils. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 515-69-5
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.372
MDL Number MFCD03846910
InChI Key RGZSQWQPBWRIAQ-LSDHHAIUSA-N
Synonym bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260
PubChem CID 1549992
IUPAC Name (2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
SMILES CC1=CCC(CC1)C(C)(CCC=C(C)C)O

Specifications

Density 0.93
Flash Point 135°C (275°F)
Refractive Index 1.495
Quantity 50 g
Beilstein 5733954
Sensitivity Air Sensitive
Merck Index 14,1241
Solubility Information Soluble in alcohol,natural,mineral and synthetic oils. Insoluble in water.
Formula Weight 222.37
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material alpha-Bisabolol
Show More Show Less
EINECSNumber 208-205-9
RTECSNumber MJ9685000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only