missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butene, 2M solution in THF, AcroSeal™

Catalog No. AC426391000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426391000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426391000 Supplier Thermo Scientific Chemicals Supplier No. 426391000

missing translation for 'validMainframeMsg'

CAS: 513-35-9 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material 2-Methyl-2-butene
Name Note 2M solution in THF
CAS 513-35-9, 109-99-9
CAS Min % 16.8, 82.36
CAS Max % 17.64, 83.2
Color Colorless to Yellow
Density 0.8350g/mL
Flash Point −46°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H10
Linear Formula CH3CH=C(CH3)2
Quantity 100 mL
MDL Number MFCD00009276
Specific Gravity 0.835
Merck Index 15, 601
Synonym 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
InChI Key BKOOMYPCSUNDGP-UHFFFAOYSA-N
SMILES CC=C(C)C
IUPAC Name 2-methylbut-2-ene
Molecular Weight (g/mol) 70.14
PubChem CID 10553
ChEBI CHEBI:77916
Formula Weight 70.13
Physical Form Liquid
Show More Show Less

RUO – Research Use Only