accessibility menu, dialog, popup

UserName

CAS RN 51174-12-0

CH3H3CCH3CH3H2C

CAS RN 51174-12-0

IUPAC Name: 2,4,4-trimethylhex-1-ene
Synonyms: 2,4,4-trimethylhex-1-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: AVKVJWFFVHGHLJ-UHFFFAOYSA-N
SMILES: CCC(C)(C)CC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results