missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-hexene 97.0+%, TCI America™

Catalog No. T07785ML
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
T07785ML 5 mL
1 options

Catalog No. T07785ML

Supplier: TCI America T07785ML

Only null left

Chemical Identifiers

CAS 51174-12-0
Molecular Formula C9H18
Molecular Weight (g/mol) 126.243
MDL Number MFCD00048626
InChI Key AVKVJWFFVHGHLJ-UHFFFAOYSA-N
PubChem CID 142817
IUPAC Name 2,4,4-trimethylhex-1-ene
SMILES CCC(C)(C)CC(=C)C

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,4-Trimethyl-1-hexene
EINECSNumber (2)-0025
TSCA No