accessibility menu, dialog, popup

UserName

FNHOHNO

CAS RN 51-21-8

IUPAC Name: 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: fluorouracil|fluorouracil (inn)
Molecular Weight (g/mol): 130.08
Molecular Formula: C4H3FN2O2
InChi Key: GHASVSINZRGABV-UHFFFAOYSA-N
SMILES: FC1=CNC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (6)
Physical Form 
  • (5)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (5)
  • (1)
  • (3)