missing translation for 'onlineSavingsMsg'
Learn More

Fluorouracil Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017817
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017817 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017817 Supplier Merck Emd Millipore Supplier No. PHR1227500MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 51-21-8
Physical Form Neat
Linear Formula C4H3 FN2O2
MDL Number MFCD00006018
Melting Point 282°C to 286°C
Synonym 5-Fluorouracil; 2,4-Dihydroxy-5-fluoropyrimidine; 5-FU; 5-Fluoro-2,4(1 H,3 H)-pyrimidinedione
Type Certified Reference Material
Quantity 500 mg
Formula Weight 130.08
Grade Certified Reference Material