accessibility menu, dialog, popup

UserName

NH2NNH2NN

CAS RN 504-08-5

IUPAC Name: 1,3,5-triazine-2,4-diamine
Synonyms: 1,3,5-triazine-2,4-diamine diamino-s-triazine guanamine formoguanamine
Molecular Weight (g/mol): 111.11
Molecular Formula: C3H5N5
InChi Key: VZXTWGWHSMCWGA-UHFFFAOYSA-N
SMILES: NC1=NC(N)=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results