CAS RN 50-28-2
IUPAC Name:
(1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Synonyms:
estradiol ovahormon delta-estradiol D-3,17β-estradiol esclim menostar amnestrogen estradurin estrodiolum 雌二醇
Molecular Weight (g/mol):
272.39
Molecular Formula:
C18H24O2
InChi Key:
VOXZDWNPVJITMN-ZBRFXRBCSA-N
SMILES:
C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O
1
–
2
of
2
results
1
2
beta-Estradiol, 99% (dry wt.), ca 3% water
CAS: 50-28-2 Molecular Formula: C18H24O2 Molecular Weight (g/mol): 272.388 MDL Number: MFCD00003693 InChI Key: VOXZDWNPVJITMN-ZBRFXRBCSA-N Synonym: estradiol,beta-estradiol,17beta-estradiol,oestradiol,dihydrofolliculin,estrace,ovocyclin,progynon,dihydrotheelin,dihydroxyestrin PubChem CID: 5757 ChEBI: CHEBI:16469 IUPAC Name: (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol SMILES: CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O