accessibility menu, dialog, popup

UserName

H3C(S)(S)OH(S)(S)O(R)H3C(S)(R)HOOHO

CAS RN 50-24-8

IUPAC Name: (1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: prednisolone deltisilone flamasone ultracortene-hydrogen predate-50 prednis prednisolonum prednisolone preparation (product) delta(1)-dehydrocortisol cordrol
Molecular Weight (g/mol): 360.45
Molecular Formula: C21H28O5
InChi Key: OIGNJSKKLXVSLS-VWUMJDOOSA-N
SMILES: C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results