accessibility menu, dialog, popup

UserName

CAS RN 498-07-7

IUPAC Name: 6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
Synonyms: levoglucosan
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: TWNIBLMWSKIRAT-UHFFFAOYNA-N
SMILES: OC1C2COC(O2)C(O)C1O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)