accessibility menu, dialog, popup

UserName

CAS RN 498-00-0

IUPAC Name: 4-(hydroxymethyl)-2-methoxyphenol
Synonyms: vanillyl alcohol
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: ZENOXNGFMSCLLL-UHFFFAOYSA-N
SMILES: COC1=CC(CO)=CC=C1O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)