accessibility menu, dialog, popup

UserName

CAS RN 497-03-0

IUPAC Name: (2E)-2-methylbut-2-enal
Synonyms: trans-2-methyl-2-butenal
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: ACWQBUSCFPJUPN-HWKANZROSA-N
SMILES: C\C=C(/C)C=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)