accessibility menu, dialog, popup

UserName

CH3OHBHOOH

CAS RN 493035-82-8

IUPAC Name: (4-hydroxy-2-methylphenyl)boronic acid
Synonyms: 4-hydroxy-2-methylphenylboronic acid
Molecular Weight (g/mol): 151.96
Molecular Formula: C7H9BO3
InChi Key: OYIYNIONWDBJIF-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results