accessibility menu, dialog, popup

UserName

CAS RN 4906-24-5

IUPAC Name: 3-oxobutan-2-yl acetate
Synonyms: 3-oxobutan-2-yl acetate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: ZKPTYCJWRHHBOW-UHFFFAOYNA-N
SMILES: CC(OC(C)=O)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results