missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxy-2-butanone, 98%

Catalog No. AAL0692714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0692714 25 g
AAL0692706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0692714 Supplier Thermo Scientific Chemicals Supplier No. L0692714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4906-24-5 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4906-24-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00051782
InChI Key ZKPTYCJWRHHBOW-UHFFFAOYNA-N
Synonym 3-acetoxy-2-butanone, acetoin acetate, 2-acetoxy-3-butanone, 2-butanone, 3-acetyloxy, 3-acetyloxy-2-butanone, 2-butanon-3-ol, acetate, acetic acid 1-methyl-2-oxopropyl ester, 2-butanon-3-yl acetate, 3-hydroxy-2-butanone acetate, acetyl methyl carbinyl acetate
PubChem CID 521246
IUPAC Name 3-oxobutan-2-yl acetate
SMILES CC(OC(C)=O)C(C)=O

Specifications

Density 1.025
Boiling Point 154°C to 155°C
Flash Point 66°C (150°F)
Refractive Index 1.4135
Quantity 25 g
Beilstein 1756882
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material 3-Acetoxy-2-butanone
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only