CAS RN 489-32-7
CAS RN 489-32-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Icariin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Icariin is a major flavonoid isolated from Epimedium spp. leaves, used as a traditional Chinese medicine to nourish the kidney and reinforce yang.
3
Icariin 96.0+%, TCI America™
CAS: 489-32-7 Molecular Formula: C33H40O15 Molecular Weight (g/mol): 676.668 MDL Number: MFCD00210516 InChI Key: TZJALUIVHRYQQB-JOJMVVPMSA-N PubChem CID: 131801625 IUPAC Name: 5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one SMILES: CC1C(C(C(C(O1)OC2=C(OC3=C(C2=O)C(=CC(=C3CC=C(C)C)OC4C(C(C(C(O4)CO)O)O)O)O)C5=CC=C(C=C5)OC)O)O)O