missing translation for 'onlineSavingsMsg'
Learn More

Icariin 96.0+%, TCI America™

Catalog No. p-7131635
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0862200MG 200 mg
I08621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0862200MG Supplier TCI America Supplier No. I0862200MG
Only null left

Chemical Identifiers

CAS 489-32-7
Molecular Formula C33H40O15
Molecular Weight (g/mol) 676.668
MDL Number MFCD00210516
InChI Key TZJALUIVHRYQQB-JOJMVVPMSA-N
PubChem CID 131801625
IUPAC Name 5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OC2=C(OC3=C(C2=O)C(=CC(=C3CC=C(C)C)OC4C(C(C(C(O4)CO)O)O)O)O)C5=CC=C(C=C5)OC)O)O)O

Specifications

Melting Point 237°C
Color Yellow
Quantity 200 mg
Formula Weight 676.67
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Icariin
RTECSNumber DJ2980500
TSCA No
Recommended Storage Refrigerator