missing translation for 'onlineSavingsMsg'
Learn More

Adonitol, MP Biomedicals

Catalog No. ICN10025205
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10025205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10025205 Supplier MP Biomedicals Inc Supplier No. 0210025205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution

Specifications

CAS 488-81-3
Density 1.53g/cm3
Molecular Formula C5H12O5
Quantity 5 g
MDL Number MFCD00064291
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
SMILES C(C(C(C(CO)O)O)O)O
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 152.146
PubChem CID 6912
Formula Weight 152.15
Physical Form Powder
Chemical Name or Material Adonitol
Show More Show Less
EINECSNumber 207-685-7