CAS RN 4860-03-1
IUPAC Name:
1-chlorohexadecane
Synonyms:
1-chlorohexadecane
Molecular Weight (g/mol):
260.89
Molecular Formula:
C16H33Cl
InChi Key:
CLWAXFZCVYJLLM-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCl
1
–
2
of
2
results
1
2
1-Chlorohexadecane 98.0+%, TCI America™
CAS: 4860-03-1 Molecular Formula: C16H33Cl Molecular Weight (g/mol): 260.89 MDL Number: MFCD00000959 InChI Key: CLWAXFZCVYJLLM-UHFFFAOYSA-N Synonym: cetyl chloride,hexadecyl chloride,hexadecane, 1-chloro,n-hexadecyl chloride,chlorohexadecane,unii-wq5zks3ek3,wq5zks3ek3,1-chloro-hexadecane,1-chloranylhexadecane,dsstox_cid_7580 PubChem CID: 20993 IUPAC Name: 1-chlorohexadecane SMILES: CCCCCCCCCCCCCCCCCl