missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorohexadecane, 97%

Catalog No. AAB23406AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23406AP 500 mL
AAB23406AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB23406AP Supplier Thermo Scientific Chemicals Supplier No. B23406AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4860-03-1 | C9H7F3O3 | 220.15 g/mol

1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

Solubility
Soluble in ether. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 4860-03-1
MDL Number MFCD00000959

Specifications

Melting Point 8°C to 10°C
Density 0.865
Boiling Point 322°C
Flash Point 136°C (276°F)
Molecular Formula C16H33Cl
Refractive Index 1.449
Linear Formula CH3(CH2)15Cl
Quantity 500 mL
Beilstein 1748498
Synonym cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580
Solubility Information Soluble in ether. Insoluble in water.
InChI Key CLWAXFZCVYJLLM-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCl
IUPAC Name 1-chlorohexadecane
Molecular Weight (g/mol) 260.89
PubChem CID 20993
Formula Weight 260.89
Percent Purity 97%
Chemical Name or Material 1-Chlorohexadecane
Show More Show Less
EINECSNumber 225-461-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only