missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorohexadecane 98.0+%, TCI America™

Catalog No. C0183500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0183500ML 500 mL
C018325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0183500ML Supplier TCI America Supplier No. C0183500ML
Only null left

Chemical Identifiers

CAS 4860-03-1
Molecular Formula C16H33Cl
Molecular Weight (g/mol) 260.89
MDL Number MFCD00000959
InChI Key CLWAXFZCVYJLLM-UHFFFAOYSA-N
Synonym cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580
PubChem CID 20993
IUPAC Name 1-chlorohexadecane
SMILES CCCCCCCCCCCCCCCCCl

Specifications

Color Colorless
Boiling Point 195°C
Quantity 500 mL
Formula Weight 260.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorohexadecane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes