accessibility menu, dialog, popup

UserName

CH3OOOOH3C

CAS RN 4841-85-4

IUPAC Name: 1,2-diethyl cyclohex-4-ene-1,2-dicarboxylate
Synonyms: 1,2-diethyl cyclohex-4-ene-1,2-dicarboxylate
Molecular Weight (g/mol): 226.27
Molecular Formula: C12H18O4
InChi Key: OWVOHDKOANTMJU-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CC=CCC1C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results