missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cis-4-Cyclohexene-1,2-dicarboxylate 98.0+%, TCI America™

Catalog No. D37145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37145G Supplier TCI America Supplier No. D37145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4841-85-4
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.272
InChI Key OWVOHDKOANTMJU-AOOOYVTPSA-N
Synonym cis-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester
PubChem CID 11276195
IUPAC Name diethyl (1S,2R)-cyclohex-4-ene-1,2-dicarboxylate
SMILES CCOC(=O)C1CC=CCC1C(=O)OCC

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 226.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl cis-4-Cyclohexene-1,2-dicarboxylate
EINECSNumber (3)-2437
TSCA No