accessibility menu, dialog, popup

UserName

H3COHOOHO

CAS RN 481-74-3

IUPAC Name: 1,8-dihydroxy-3-methyl-9,10-dihydroanthracene-9,10-dione
Synonyms: turkey rhubarb C.I. natural yellow 23 chrysophanol chrysophanic acid 3-methylchrysazin chrysophansaeure
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: LQGUBLBATBMXHT-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C2C(=O)C3=C(O)C=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results