missing translation for 'onlineSavingsMsg'
Learn More

Chrysophanic Acid 96.0+%, TCI America™

Catalog No. C2505100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2505100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2505100MG Supplier TCI America Supplier No. C2505100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 481-74-3
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00001208
InChI Key LQGUBLBATBMXHT-UHFFFAOYSA-N
Synonym chrysophanol, chrysophanic acid, 3-methylchrysazin, turkey rhubarb, 1,8-dihydroxy-3-methylanthraquinone, crysophanic acid, c.i. natural yellow 23, 1,8-dihydroxy-3-methyl-9,10-anthracenedione, 4,5-dihydroxy-2-methylanthraquinone, crysophanol
PubChem CID 10208
ChEBI CHEBI:3687
IUPAC Name 1,8-dihydroxy-3-methyl-9,10-dihydroanthracene-9,10-dione
SMILES CC1=CC(O)=C2C(=O)C3=C(O)C=CC=C3C(=O)C2=C1

Specifications

Melting Point 194°C
Quantity 100 mg
Formula Weight 254.24
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Chrysophanic Acid
RTECSNumber CB6725000
TSCA No