accessibility menu, dialog, popup

UserName

BOOCH3HOOH

CAS RN 480424-49-5

IUPAC Name: (3-formyl-2-methoxyphenyl)boronic acid
Synonyms: 3-formyl-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: DUROSIJIMLRFHR-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1C=O)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results