missing translation for 'onlineSavingsMsg'
Learn More

3-Formyl-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10881G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10881G Supplier TCI America Supplier No. F10881G

Specifications

Chemical Name or Material 3-Formyl-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 480424-49-5
Melting Point 128°C
Color White-Yellow
Molecular Formula C8H9BO4
Quantity 1 g
MDL Number MFCD08689486
Synonym 3-formyl-2-methoxyphenyl boronic acid, 3-formyl-2-methoxybenzeneboronic acid, boronic acid, b-3-formyl-2-methoxyphenyl, acmc-209kbb, 3-formyl-2-methoxyphenylboronicacid, 3-formyl-2-methoxy-phenyl boronic acid, 3-methanoyl-2-methoxy-phenyl boronic acid, boronic acid,b-3-formyl-2-methoxyphenyl, 3-formyl-2-methoxyphenylboronic acid
InChI Key DUROSIJIMLRFHR-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)C=O)OC)(O)O
IUPAC Name (3-formyl-2-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 179.966
PubChem CID 16218167
Formula Weight 179.97
Physical Form Crystalline Powder
Show More Show Less