accessibility menu, dialog, popup

UserName

CAS RN 479-59-4

IUPAC Name: 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
Synonyms: julolidine
Molecular Weight (g/mol): 173.26
Molecular Formula: C12H15N
InChi Key: DZFWNZJKBJOGFQ-UHFFFAOYSA-N
SMILES: C1CN2CCCC3=C2C(C1)=CC=C3
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)