missing translation for 'onlineSavingsMsg'
Learn More

Julolidine, 98%

Catalog No. AAB2297614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2297614 25 g
AAB2297603 1 g
AAB2297606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2297614 Supplier Thermo Scientific Chemicals Supplier No. B2297614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 479-59-4 | C12H15N | 173.259 g/mol

Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.

Chemical Identifiers

CAS 479-59-4
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
MDL Number MFCD00006917
InChI Key DZFWNZJKBJOGFQ-UHFFFAOYSA-N
Synonym julolidine, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine, 1h,5h-benzo ij quinolizine, 2,3,6,7-tetrahydro, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline, 2,3,6,7-tetrahydro-1h,5h-benzo i,j quinolizine, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline, 2,6,7-tetrahydro-1h,5h-benzo ij quinolizine
PubChem CID 68069
SMILES C1CC2=C3C(=CC=C2)CCCN3C1

Specifications

Melting Point 32°C to 37°C
Boiling Point 170°C to 173°C (31 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 139925
Sensitivity Air sensitive
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 173.26
Percent Purity 98%
Chemical Name or Material Julolidine
EINECSNumber 207-535-0
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only