CAS RN 475250-52-3
CAS RN 475250-52-3
IUPAC Name:
2-[(4-methoxyphenyl)methyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-[(4-methoxyphenyl)methyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
248.13
Molecular Formula:
C14H21BO3
InChi Key:
HPNLRRQVSZVKHY-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CB2OC(C)(C)C(C)(C)O2)C=C1
1
–
1
of
1
results
1
2-(4-Methoxybenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™
CAS: 475250-52-3 Molecular Formula: C14H21BO3 Molecular Weight (g/mol): 248.129 MDL Number: MFCD10698526 InChI Key: HPNLRRQVSZVKHY-UHFFFAOYSA-N Synonym: 4-Methoxybenzylboronic Acid Pinacol Ester PubChem CID: 12012759 IUPAC Name: 2-[(4-methoxyphenyl)methyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)CC2=CC=C(C=C2)OC